Order Entry
Export
Orders LinkContactUsLinkComponent
 

Evaporating dishes, high shape

500 ml, 55 mm, 110 mm
Catalog # BOCH8562
Supplier:   BOCHEM
Evaporating dishes, high shape
Catalog # BOCH8562
Supplier:  BOCHEM
Evaporating dishes, high shape
Evaporating dishes, high shape

500 ml, 55 mm, 110 mm

Price
50,00 €
Log in for availability
1 * 1 items
Product Details & Documents

Evaporating dish, stainless steel, with contact bottom for heating plates and magnetic stirrers.

  • Very good temperature resistance
  • Good chemical resistance
  • Easy to clean
Specifications
  • Ext.Ø:
    110 mm
  • Capacity:
    500 ml
  • Height:
    55 mm