Pipette tips, ART™
ART™ 20 SoftFit®
Catalog # 732-1522
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Tip pipette 20µl HLT 20L SoftFit-L reinforced tray LTS series hinged rack non-sterile 1 * 960 items Catalog #613-1870
Product 613-1870 is similar to 732-1522 but manufacturer and specifications may vary.
Product Details & Documents
Transparent PP, sterile or non sterile.
- Available as non sterile bulk or racked; and as sterile racked
- Certified sterile tips, free of DNase, RNase, DNA and pyrogens (sterilised by gamma radiation)
- SoftFit® design assures a consistent, reliable fit for all channels of the pipette, which in turn ensures reliable distribution of the liquid
Ordering information: Supplied loose bagged or in racks.
Specifications
- Tip style:ART™ 20 SoftFit®