Erlenmeyer flasks, disposable, with screw cap, Nalgene®
500 ml, 45-430
Catalog # 214-0138
Supplier: Thermo Fisher Scientific
Erlenmeyer flasks, disposable, with screw cap, Nalgene®
Catalog # 214-0138
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
PETG, transparent, with HDPE screw cap.
- Leakproof HDPE screw cap closure can be opened partially for venting by twisting a quarter turn
- Packaged individually for easy storage and handling
- Reduced risk of cross-contamination
Sterile, disposable Erlenmeyer flasks reduce the risk of cross-contamination. Ideal for shakers and suspension cultures, and for creating or storing media. Raised graduations for approximate reading of the contents.
Specifications
- Thread:45-430
- Capacity:500 ml
- Cat. No.:214-0138
- Environmental certification:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact