Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Extractors
Automatic Extractor Systems
Accessories for extractors
Accessories for extractors
Print
Accessories for extractors
Solvent beaker, standard
Catalog # BUCH037276
Supplier:
BUCHI
Accessories for extractors
Catalog # BUCH037276
Supplier:
BUCHI
Expand image
Return to Top
Accessories for extractors
Solvent beaker, standard
Specifications
Add to cart
Jump to:
Specifications
Price
401,00 €
Log in
for availability
Unit of Measure
1 * 4 items
-
+
Add to cart
Other Product Family Options Available
Return Policy
Product Details & Documents
Specifications
Description:
Solvent beaker, standard
Seq:
02