Order Entry
Export
Orders LinkContactUsLinkComponent
Sarcosine

Sarcosine

Glass bottle for solids, 25 g
Catalog # BMOL54687-1
Sarcosine
Catalog # BMOL54687-1
CAS Number:  107-97-1
Sarcosine

Glass bottle for solids, 25 g

CAS Number:  107-97-1
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  H-Sar-OH, N-Methylglycine, Methylaminoacetic acid
  • CAS Number:107-97-1
  • Formula:C₃H₇NO₂
  • MDL Number:MFCD00004279
  • Molecular Weight:89.09 g/mol
  • Merck Index:12,08519
  • Boiling Pt:185 °C (1013 hPa)
  • Melting Pt:63 °C
  • Density:1.301 g/cm³ (20 °C)
  • EINECS:203-538-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications