Order Entry
Export
Orders LinkContactUsLinkComponent
 

Buffer, TRIS-glycine-SDS solution 10X concentrate (TGS buffer)

TRIS-glycine-SDS buffer solution 10X concentrate (TGS buffer), Glass bottle, 1 L
Catalog # BMOL54662-2
Buffer, TRIS-glycine-SDS solution 10X concentrate (TGS buffer)
Catalog # BMOL54662-2
CAS Number:  
Buffer, TRIS-glycine-SDS solution 10X concentrate (TGS buffer)

TRIS-glycine-SDS buffer solution 10X concentrate (TGS buffer), Glass bottle, 1 L

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 L
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Description:
    TRIS-glycine-SDS buffer solution 10X concentrate (TGS buffer)

Specifications