Buffer, TRIS-glycine-SDS solution 10X concentrate (TGS buffer)
TRIS-glycine-SDS buffer solution 10X concentrate (TGS buffer), Glass bottle, 500 ml
Catalog # BMOL54662-1
Supplier: BIOMOL RESEARCH LABORATORIES
Buffer, TRIS-glycine-SDS solution 10X concentrate (TGS buffer)
Catalog # BMOL54662-1
Supplier: BIOMOL RESEARCH LABORATORIES
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:500 ml
- Description:TRIS-glycine-SDS buffer solution 10X concentrate (TGS buffer)



