n-Decyl-β-D-maltoside
Glass bottle for solids, 5 g
Catalog # BMOL50165-2
Supplier: BIOMOL RESEARCH LABORATORIES
n-Decyl-β-D-maltoside
Catalog # BMOL50165-2
Supplier: BIOMOL RESEARCH LABORATORIES
CAS Number: 82494-09-5
Jump to:
Product Details & Documents
Synonyms: DM, N-Decyl-Beta-Maltoside, Decyl 4-O-a-D-glucopyranosyl-b-D-glucopyranoside, n-Decyl-beta-D-maltopyranoside, Decyl b-D-maltopyranoside
- CAS Number:82494-09-5
- Formula:C₂₂H₄₂O₁₁
- MDL Number:MFCD00061624
- Molecular Weight:482.56 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



