L(-)-Tyrosine
Glass bottle for solids, 1 kg
Catalog # BMOL08125-2
Supplier: BIOMOL RESEARCH LABORATORIES
Jump to:
Product Details & Documents
Synonyms: L-Tyrosine, (S)-(-)-Tyrosine, Tyrosine, H-Tyr-OH
- CAS Number:60-18-4
- Formula:C₉H₁₁NO₃
- MDL Number:MFCD00002606
- Molecular Weight:181.19 g/mol
- Merck Index:13,09907
- Boiling Pt:351 °C (1013 hPa)
- Melting Pt:285 °C
- Density:1.41 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-460-4
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg



