Order Entry
Export
Orders LinkContactUsLinkComponent
Taurine

Taurine

Glass bottle for solids, 500 g
Catalog # BMOL07579-2
Taurine
Catalog # BMOL07579-2
CAS Number:  107-35-7
Taurine

Glass bottle for solids, 500 g

CAS Number:  107-35-7
Price
Inquire For Price
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  2-Aminoethanesulphonic acid
  • CAS Number:107-35-7
  • Formula:C₂H₇NO₃S
  • MDL Number:MFCD00008197
  • Molecular Weight:125.15 g/mol
  • Merck Index:13,09163
  • Melting Pt:300 °C
  • Storage Temperature:Ambient
  • EINECS:203-483-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications