L-Methionine
Glass bottle for solids, 100 g
Catalog # BMOL06023-1
Supplier: BIOMOL RESEARCH LABORATORIES
Jump to:
Product Details & Documents
Synonyms: (S)-(-)-2-Amino-4-(methylthio)butyric acid, (S)-(-)-Methionine, L(-)-Methionine, (S)-(-)-2-Amino-4-(methylthio)butanoic acid, H-Met-OH, Methionine, (S)-2-Amino-4-(methylthio)butanoic acid
- CAS Number:63-68-3
- Formula:C₅H₁₁NO₂S
- MDL Number:MFCD00063097
- Molecular Weight:149.21 g/mol
- Merck Index:13,06004
- Boiling Pt:278 °C (1013 hPa)
- Melting Pt:136 °C
- Density:1.373 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-562-9
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



