D-(+)-Maltose monohydrate
Glass bottle for solids, 500 g
Catalog # BMOL05957-2
Supplier: BIOMOL RESEARCH LABORATORIES
D-(+)-Maltose monohydrate
Catalog # BMOL05957-2
Supplier: BIOMOL RESEARCH LABORATORIES
CAS Number: 6363-53-7
Jump to:
Product Details & Documents
Synonyms: Maltobiose hydrate, Malt Sugar, beta-Maltose monohydrate, Maltose monohydrate, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal hydrate, 4-O-α-D-Glucopyranosyl-D-glucose hydrate
- CAS Number:6363-53-7
- Formula:C₁₂H₂₂O₁₁·1H₂O
- MDL Number:MFCD00149343
- Molecular Weight:360.32 g/mol
- Merck Index:13,05736
- Melting Pt:160…165 °C
- Storage Temperature:Ambient
- EINECS:200-716-5
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g



