L(+)-Glutamic acid
Glass bottle for solids, 500 g
BMOL04750-1
: (S)-(+)-2-Aminopentanedioic acid, L(+)-1-Aminopropane-1,3-dicarboxylic acid, (S)-(+)-α-Glutamic acid, H-Glu-OH, L-Glutamic acid, L(+)-2-Aminopentanedioic acid, (S)-(+)-Glutamic acid, (S)-(+)-1-Aminopropane-1,3-dicarboxylic acid, L(+)-α-Glutamic acid
- :56-86-0
- :HO₂CCH₂CH₂CH(NH₂)CO₂H
- :MFCD00002634
- :147.13 g/mol
- :13,04482
- :325 °C (1013 hPa)
- :160 °C
- :1.538 g/cm³ (20 °C)
- :Ambient
- :200-293-7
- Pack type:Glass bottle for solids
- Pk:500 g