Orders LinkContactUsLinkComponent
Ring stand for viscosity flow cups
Ring stand for viscosity flow cups
Viscosity convertor of flow time to viscosity
Catalog # 769-1151
Supplier:   Industrial Physics
Ring stand for viscosity flow cups
Catalog # 769-1151
Supplier:  Industrial Physics
Ring stand for viscosity flow cups
Ring stand for viscosity flow cups
Viscosity convertor of flow time to viscosity
Price
87,50 €
Log in for availability
1 * 1 items
Product Details & Documents
Galvanised steel simple ring stand suitable for DIN, ISO and ASTM viscosity flow cups.
Documents
Specifications
  • Description:
    Viscosity convertor of flow time to viscosity
  • Seq:
    01