Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Reagenzresthärte,Typ 310, VE=500 ml 1 * 500 mL
Catalog # 713-1301
Supplier:
Heyl GmbH & Co, Gebr.
Expand image
Reagenzresthärte,Typ 310, VE=500 ml 1 * 500 mL
Catalog # 713-1301
Supplier:
Heyl GmbH & Co, Gebr.