Order Entry
Export
Orders LinkContactUsLinkComponent
Platinum/iridium wire

Platinum/iridium wire

Platinum/iridium wire, Ø 0,8 mm
Catalog # 631-7116
Supplier:   CHEMPUR
Platinum/iridium wire
Catalog # 631-7116
Supplier:  CHEMPUR
Platinum/iridium wire

Platinum/iridium wire, Ø 0,8 mm

Price
Inquire For Price
Log in for availability
1 * 1 cm
Product Details & Documents

Platinum/Iridium 90/10 wire.

Platinum wire is used in many applications including gasket or o-ring fabrication, thermocouples, electrical contacts, loops and heater coils.

Specifications
  • Description:
    Platinum/iridium wire, Ø 0,8 mm
  • Seq:
    01