Order Entry
Export
Orders LinkContactUsLinkComponent
Counting chambers, Imp. Neubauer

Counting chambers, Imp. Neubauer

Counting chamber, double, Imp. Neubauer, 0,1 mm
Catalog # 631-1111
Supplier:   LABOR OPTIK
Counting chambers, Imp. Neubauer
Catalog # 631-1111
Supplier:  LABOR OPTIK
Counting chambers, Imp. Neubauer
Counting chambers, Imp. Neubauer

Counting chamber, double, Imp. Neubauer, 0,1 mm

Price
58,30 €
Log in for availability
1 * 1 items
Product Details & Documents

For counting leucocytes, thrombocytes and erythrocytes. All group squares have triple border lines.

  • Four large squares in the corners are divided into 16 squares each (leucocyte counting)
  • Large square in the middle divided in 25 squares, each of which is further divided into 16 small squares
  • The middle line determines whether cells in the border area are to be included in one square or the other

Counting grid: 9

Large squares: 1 mm² each

Large squares: 1 mm² each

Documents

Specifications
  • Depth of chamber:
    0,1 mm
  • Description:
    Counting chamber, double, Imp. Neubauer