Orders LinkContactUsLinkComponent
Penetration probes, K-Type
Penetration probes, K-Type
Spring-loaded V4A tube with Ø 3 mm needle-shaped insertion tip, with plastic handle, anti-buckling glanding, 1 m silicone cable and miniature flat plug
Response time (T90): water 0,4 m/s ~5 s, −65…+550 °C
Catalog # 620-4708
Supplier:   Greisinger
Penetration probes, K-Type
Catalog # 620-4708
Supplier:  Greisinger
Penetration probes, K-Type
Penetration probes, K-Type
Spring-loaded V4A tube with Ø 3 mm needle-shaped insertion tip, with plastic handle, anti-buckling glanding, 1 m silicone cable and miniature flat plug
Response time (T90): water 0,4 m/s ~5 s, −65…+550 °C
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Range:
    −65…+550 °C
  • Description:
    Spring-loaded V4A tube with Ø 3 mm needle-shaped insertion tip, with plastic handle, anti-buckling glanding, 1 m silicone cable and miniature flat plug
    Response time (T90): water 0,4 m/s ~5 s
  • Seq:
    03