Order Entry
Export
Orders LinkContactUsLinkComponent
Cleaning brushes for butyrometer

Cleaning brushes for butyrometer

Cleaning brush for butyrometer body, Wire, 270 mm
Catalog # 614-2036
Supplier:   GERBER FUNKE
Cleaning brushes for butyrometer
Catalog # 614-2036
Supplier:  GERBER FUNKE
Cleaning brushes for butyrometer

Cleaning brush for butyrometer body, Wire, 270 mm

Price
2,30 €
Log in for availability
1 * 1 items
Product Details & Documents

Cleaning brush for butyrometer body

  • Total length: 270 mm
  • Brush length: 100 mm
Specifications
  • Handle material:
    Wire
  • Description:
    Cleaning brush for butyrometer body
  • Seq:
    01
  • Total length:
    270 mm