Standard microliter syringes
500 µl, 1
Catalog # 613-1720
Supplier: Hospidex
Jump to:
Product Details & Documents
Providing ultra-precise analyses. The plunger extends down into the tip of the needle ensuring that no dead volume is created.
The bevelled needle tip is 50 mm long.
Specifications
- Type of tip:1
- Volume:500 µl