Orders LinkContactUsLinkComponent
Pipette containers, SPECICON
Rack, GESTICON, stainless steel, for 4 round or rectangular pipette containers (up to 70×70 mm), Dimensions 195×229×235 mm
Catalog # 612-1865
Supplier:   schuett-biotec GmbH
Pipette containers, SPECICON
Catalog # 612-1865
Pipette containers, SPECICON
Rack, GESTICON, stainless steel, for 4 round or rectangular pipette containers (up to 70×70 mm), Dimensions 195×229×235 mm
Price
114,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Containers are made of thick walled aluminium, anodised in gold colour.
  • With sterilisation opening and bayonet closures
  • Can be hermetically sealed
They can be used for the sterilisation and storage of pipettes.
Specifications
  • Description:
    Rack, GESTICON, stainless steel, for 4 round or rectangular pipette containers (up to 70×70 mm), Dimensions 195×229×235 mm
  • Seq:
    03
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Silicone insert (2 mm thick), prevents damage to pipettes caused by sudden shock, for pipette containers VARICON RS (lower part) and SPECICON (lower and upper part)
    Silicone insert (2 mm thick), prevents damage to pipettes caused by sudden shock, for pipette containers VARICON RS (lower part) and SPECICON (lower and upper part)
    SCUT3523213
    Log in for availability
    1 * 1 items - 5,90 €