Glass Adapters
ROBU® Glass Adapter for Crucibles is designed to securely connect filter crucibles to other laboratory apparatus. Made from durable glass, it ensures a leak-free fit, offering reliable and efficient performance during filtration and chemical processes.
- Leak-free connection
- Durable glass construction
- Resistant to heat and chemicals
- Ensures secure attachment for crucibles
- Compatible with various filtration setups
- Easy to clean and reusable
- Reliable performance in laboratory filtration processes
The ROBU Glass Adapter for Crucibles is designed to provide a secure and leak-free connection between filter crucibles and other laboratory apparatus. Constructed from durable, heat-resistant glass, this adapter is highly resistant to chemicals and ensures consistent, reliable performance during filtration processes. It is compatible with a wide range of filtration setups, offering versatility and ease of use in various laboratory applications. The adapter is easy to clean and reusable, making it a cost-effective solution for maintaining high standards in laboratory filtration work.
: In compliance with ISO 4796-80 and ASTM E128-99 standards.
Documents
- Ext.Ø:10 mm
- Description:Glass adapter; inner tube Ø: 50 mm; outer tube Ø: 10 mm
- Material:Borosilicate glass 3.3
- Seq:4



