Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
AF20 Ice flaker/maker s/steel115kg/day 70kg bin storage 1 * 1 items
Catalog # 474-1405
Supplier:
SCOTSMAN
Expand image
AF20 Ice flaker/maker s/steel115kg/day 70kg bin storage 1 * 1 items
Catalog # 474-1405
Supplier:
SCOTSMAN