Orders LinkContactUsLinkComponent
Boiling Stones
Boiling Stones
Boiling stones, grade B, for analytical work
Catalog # 459-9122
Supplier:   POLLATH
Boiling Stones
Catalog # 459-9122
Supplier:  POLLATH
Boiling Stones
Boiling Stones
Boiling stones, grade B, for analytical work
Price
24,40 €
Log in for availability
1 * 200 g
Product Details & Documents
Grade A: For preparative work. Can be used in aqueous, neutral, basic media. Highly effective even in the case of multiple interruptions of the boiling process in vacuum distillation.
Grade B: For analytical work, with acidic, neutral, basic media. Jump after interruption again. Not suitable for vacuum distillation.
Specifications
  • Description:
    Boiling stones, grade B, for analytical work