Order Entry
Export
Orders LinkContactUsLinkComponent
Heating tapes, TeMS2

Heating tapes, TeMS2

Heating tape TeMS6, 61 m length, 160 W
Catalog # 454-2191
Supplier:   ISOPAD
Heating tapes, TeMS2
Catalog # 454-2191
Supplier:  ISOPAD
Heating tapes, TeMS2
Heating tapes, TeMS2

Heating tape TeMS6, 61 m length, 160 W

Price
327,20 €
Log in for availability
1 * 1 items
Product Details & Documents

Isopad IT-TeMS is a preterminated and PTFE insulated flexible heating tape.

  • Temperature resistant up to 260 °C
  • Non hazardous and used in ordinary areas
  • Parallel arrangement ensures a wide heat transfer surface

IT-TeMS2 is used in applications ranging from simple frost protection to temperature maintenance on short pipework and apparatus as well as for general container heating applications. TeMS4 or TeMS6 tapes can be used when more power is required.

Specifications
  • Description:
    Heating tape TeMS6, 61 m length, 160 W
  • Seq:
    09