Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Base scaffold foot thumbscrew clamping accepts diameters up to 13mm 1 * 1 items
Catalog # 241-2121
Supplier:
HARVARD APPARATUS
Expand image
Base scaffold foot thumbscrew clamping accepts diameters up to 13mm 1 * 1 items
Catalog # 241-2121
Supplier:
HARVARD APPARATUS