Order Entry
Export
Orders LinkContactUsLinkComponent
 

Retort rods, unthreaded

13 mm, 750 mm
  241-2115
Retort rods, unthreaded
  241-2115
Retort rods, unthreaded

13 mm, 750 mm

 
43,80 €
   
1 * 1 items
 

Stainless steel.

  • Brilliantly polished stainless steel with good surface hardness that will not score from clamp binding screws
  • Hollow, lightweight rods (Ø 13 mm) with continuously welded walls (1,6 mm thick)
  • Good chemical resistance
  • Red caps
 
  • Ø:
    13 mm
  • Length:
    750 mm
  • Seq:
    05