Orders LinkContactUsLinkComponent
Connectors, PP
Connector, straight, female luer lock/tubing nipple, 2,4 mm
Catalog # 229-0526
Supplier:   PIEPER FILTER
Connectors, PP
Catalog # 229-0526
Supplier:  PIEPER FILTER
Connectors, PP
Connectors, PP
Connector, straight, female luer lock/tubing nipple, 2,4 mm
Price
22,00 €
Log in for availability
1 * 10 items
Product Details & Documents
Connectors, made of PP, natural.
  • Good chemical resistance
  • Good temperature resistance
Specifications
  • Tubing int.Ø:
    2,4 mm
  • Description:
    Connector, straight, female luer lock/tubing nipple
  • Seq:
    08