Connectors, PP
Connector, straight, male luer lock/tubing nipple, 4,0 mm
Catalog # 229-0518
Supplier: PIEPER FILTER
Jump to:
Product Details & Documents
Connectors, made of PP, natural.
- Good chemical resistance
- Good temperature resistance
Documents
Specifications
- Tubing int.Ø:4,0 mm
- Description:Connector, straight, male luer lock/tubing nipple
- Seq:04