Connectors, PP
Connector, straight, male luer lock/tubing nipple, 1,6 mm
Catalog # 229-0515
Supplier: PIEPER FILTER
Jump to:
Product Details & Documents
Connectors, made of PP, natural.
- Good chemical resistance
- Good temperature resistance
Documents
Specifications
- Tubing int.Ø:1,6 mm
- Description:Connector, straight, male luer lock/tubing nipple
- Seq:01