Order Entry
Export
Orders LinkContactUsLinkComponent
 

Conical cork stoppers

27,0 mm, 28,0 mm, 32,0 mm
Catalog # 217-9645
Conical cork stoppers
Catalog # 217-9645
Supplier:  
Conical cork stoppers

27,0 mm, 28,0 mm, 32,0 mm

Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Product Details & Documents

These conical stoppers are made of cork.

Specifications
  • Ø bottom:
    28,0 mm
  • Ø top:
    32,0 mm
  • Height:
    27,0 mm
  • Seq:
    02