Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Stoppers
PE/LDPE/HDPE Stoppers
Stoppers, PE
Stoppers, PE
Print
Stoppers, PE
Red, 14/17
Catalog # 217-8107
Supplier:
GML ALFAPLAST
Stoppers, PE
Catalog # 217-8107
Supplier:
GML ALFAPLAST
Expand image
Return to Top
Stoppers, PE
Red, 14/17
Specifications
Add to cart
Jump to:
Specifications
Price
31,10 €
Log in
for availability
Unit of Measure
1 * 1.000 items
-
+
Add to cart
Other Product Family Options Available
Return Policy
Product Details & Documents
Made of PE.
Fits glass or plastic tubes
Specifications
NS:
14/17
Colour:
Red
Seq:
01