0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Stoppers
PE/LDPE/HDPE Stoppers
Stoppers, PE
Stoppers, PE
Blue, 11/14
217-8103
:
GML ALFAPLAST
Stoppers, PE
217-8103
:
GML ALFAPLAST
10
Stoppers, PE
Blue, 11/14
26,10 €
1 * 1.000 items
-
+
Made of PE.
Fits glass or plastic tubes
NS:
11/14
Colour:
Blue
Seq:
06