Order Entry
Export
Orders LinkContactUsLinkComponent
Ice packs

Ice packs

190×140×25 mm, 1361 g
Catalog # 216-7012
Supplier:   HONIGBERG
Ice packs
Catalog # 216-7012
Supplier:  HONIGBERG
Ice packs
Ice packs

190×140×25 mm, 1361 g

Price
118,00 €
Log in for availability
1 * 6 items
Product Details & Documents

Heavy, metal-coated PE film filled with special gel cooling pad

  • Permanently leak-proof and puncture resistant
  • Content after thawing gel
  • Reusable, non-toxic

These durable and convient ice packs are designed for many applications like cooling, shipping and cold storage.

Note: Those -23 °C formulation gel packs can be placed in an insulated container at temperatures between 21 and 23 °C to keep the materials at or below 0 °C for over 40 hours.

Specifications
  • W×D×H:
    190×140×25 mm
  • Weight:
    1361 g