Storage Bins, PS
Storage bin, size 4, Green, 200×336×150 mm
Catalog # 216-3732
Supplier: hünersdorff GmbH
Jump to:
Product Details & Documents
Storage bins are particularly suitable for a wide range of applications in workshop, store, household and office.
- High load capacity
- Smooth bottom makes it suitable for transport
- Stackable longitudinally and transversely
- Made of PS, coloured
Specifications
- Colour:Green
- Description:Storage bin, size 4
- Seq:14
- W×D×H:200×336×150 mm



