Order Entry
Export
Orders LinkContactUsLinkComponent
Storage Bins, PS

Storage Bins, PS

Storage bin, size 4, Red, 200×336×150 mm
Catalog # 216-3730
Supplier:   hünersdorff GmbH
Storage Bins, PS
Catalog # 216-3730
Supplier:  hünersdorff GmbH
Storage Bins, PS
Storage Bins, PS

Storage bin, size 4, Red, 200×336×150 mm

Price
31,90 €
Log in for availability
1 * 1 items
Product Details & Documents

Storage bins are particularly suitable for a wide range of applications in workshop, store, household and office.

  • High load capacity
  • Smooth bottom makes it suitable for transport
  • Stackable longitudinally and transversely
  • Made of PS, coloured
Specifications
  • Colour:
    Red
  • Description:
    Storage bin, size 4
  • Seq:
    11
  • W×D×H:
    200×336×150 mm