Order Entry
Export
Orders LinkContactUsLinkComponent
 

Storage Bins, PS

Storage bin, size 3, Blue, 145×232×125 mm
Catalog # 216-3721
Supplier:   hünersdorff GmbH
Storage Bins, PS
Catalog # 216-3721
Supplier:  hünersdorff GmbH
Storage Bins, PS
Storage Bins, PS

Storage bin, size 3, Blue, 145×232×125 mm

Price
17,10 €
Log in for availability
1 * 1 items
Product Details & Documents

Storage bins are particularly suitable for a wide range of applications in workshop, store, household and office.

  • High load capacity
  • Smooth bottom makes it suitable for transport
  • Stackable longitudinally and transversely
  • Made of PS, coloured
Specifications
  • Colour:
    Blue
  • Description:
    Storage bin, size 3
  • Seq:
    08
  • W×D×H:
    145×232×125 mm