Storage Bins, PS
Storage bin, size 3, Red, 145×232×125 mm
Catalog # 216-3720
Supplier: hünersdorff GmbH
Jump to:
Product Details & Documents
Storage bins are particularly suitable for a wide range of applications in workshop, store, household and office.
- High load capacity
- Smooth bottom makes it suitable for transport
- Stackable longitudinally and transversely
- Made of PS, coloured
Specifications
- Colour:Red
- Description:Storage bin, size 3
- Seq:06
- W×D×H:145×232×125 mm



