Order Entry
Export
Orders LinkContactUsLinkComponent
Storage Bins, PS

Storage Bins, PS

Storage bin, size 2, Blue, 100×165×75 mm
Catalog # 216-3711
Supplier:   hünersdorff GmbH
Storage Bins, PS
Catalog # 216-3711
Supplier:  hünersdorff GmbH
Storage Bins, PS
Storage Bins, PS

Storage bin, size 2, Blue, 100×165×75 mm

Price
3,10 €
Log in for availability
1 * 1 items
Product Details & Documents

Storage bins are particularly suitable for a wide range of applications in workshop, store, household and office.

  • High load capacity
  • Smooth bottom makes it suitable for transport
  • Stackable longitudinally and transversely
  • Made of PS, coloured
Specifications
  • Colour:
    Blue
  • Description:
    Storage bin, size 2
  • Seq:
    03
  • W×D×H:
    100×165×75 mm