Order Entry
Export
Orders LinkContactUsLinkComponent
 

Storage Bins, PS

Storage bin, size 2, Red, 100×165×75 mm
  216-3710
Storage Bins, PS
  216-3710
Storage Bins, PS
Storage Bins, PS

Storage bin, size 2, Red, 100×165×75 mm

 
3,10 €
   
1 * 1 items
 

Storage bins are particularly suitable for a wide range of applications in workshop, store, household and office.

  • High load capacity
  • Smooth bottom makes it suitable for transport
  • Stackable longitudinally and transversely
  • Made of PS, coloured
 
  • Colour:
    Red
  • Description:
    Storage bin, size 2
  • Seq:
    01
  • W×D×H:
    100×165×75 mm