Order Entry
Export
Orders LinkContactUsLinkComponent
Transport and Storage Boxes

Transport and Storage Boxes

60 l, Box, storage and transport, 550×350×310 mm
Catalog # 216-2122
Supplier:   ZARGES LEICHTBAU
Transport and Storage Boxes
Catalog # 216-2122
Supplier:  ZARGES LEICHTBAU
Transport and Storage Boxes
Transport and Storage Boxes

60 l, Box, storage and transport, 550×350×310 mm

Price
388,00 €
Log in for availability
1 * 1 items
Product Details & Documents

Aluminium alloy.

  • Lid with stainless steel strap hinge, two straps and all-round seal
  • Flip lock latches, secured with anti-tamper seals or padlock
  • Flip handles on the front surfaces

Secure transportation, practical storage and protective packaging in one. The transported or stored item is securely packed and protected against external influences. Sturdy edge, lid and base profile. Stackable.

Specifications
  • Capacity:
    60 l
  • Description:
    Box, storage and transport
  • Seq:
    02
  • W×D×H int.:
    550×350×310 mm