Order Entry
Export
Orders LinkContactUsLinkComponent
Sample bottles with screw cap

Sample bottles with screw cap

2,80 ml, 13×40 mm, Yes
Catalog # 215-8341
Supplier:   MUELLER KREMPEL
Sample bottles with screw cap
Catalog # 215-8341
Supplier:  MUELLER KREMPEL
Sample bottles with screw cap
Sample bottles with screw cap

2,80 ml, 13×40 mm, Yes

Price
428,00 €
Log in for availability
1 * 530 items
Product Details & Documents

Vials made from AR glass, clear with black screw cap. Ideal for taking, storing and transporting samples.

  • Thin-walled, cylindrical
  • Tightly sealing screw cap
  • Designed for use in applications that do not require resistance to temperature
Specifications
  • Cap included:
    Yes
  • Ø×H:
    13×40 mm
  • Capacity:
    2,80 ml