Order Entry
Export
Orders LinkContactUsLinkComponent
 

Racks for test tubes

18 mm, 48 (4×12), 320×110×80 mm
  212-6345
Racks for test tubes
  212-6345
Racks for test tubes
Racks for test tubes

18 mm, 48 (4×12), 320×110×80 mm

 
231,00 €
   
1 * 1 items
 

Rack for test tubes, made from clear acrylic, constructed in two tiers.

  • Good thermal resistance
  • Rugged stable design
  • Easy to clean
 
  • Hole Ø:
    18 mm
  • No. of Holes:
    48 (4×12)
  • Seq:
    06
  • W×D×H:
    320×110×80 mm