Racks for test tubes
18 mm, 20 (2×10), 270×60×80 mm
212-6336
Jump to:
Rack for test tubes, made from clear acrylic, constructed in two tiers.
- Good thermal resistance
- Rugged stable design
- Easy to clean
Specifications
- Hole Ø:18 mm
- No. of Holes:20 (2×10)
- Seq:04
- W×D×H:270×60×80 mm



