Orders LinkContactUsLinkComponent
Labels, hazardous substance
Labels, hazardous substance
Hazard labels, 52×52 mm
Catalog # 116-1572
Supplier:   LAB IN
Labels, hazardous substance
Catalog # 116-1572
Supplier:  LAB IN
Labels, hazardous substance
Hazard labels, 52×52 mm
Price
6,70 €
Log in for availability
1 * 5 items
Product Details & Documents
Hazardous substance labels from self- adhesive foil for containers.
  • Available in three dimensions
Acc. GefStoffV.
Specifications
  • Description:
    Hazard labels
  • Dimensions:
    52×52 mm
  • Seq:
    33