Order Entry
Export
Orders LinkContactUsLinkComponent
 

Cleaning wipe rolls

Cleaning cloth Multiclean® plus, blue, Blue, 180 m × 380 mm, 2 rolls of 500 sheets
Cleaning wipe rolls
  115-2720
Cleaning wipe rolls
Cleaning wipe rolls

Cleaning cloth Multiclean® plus, blue, Blue, 180 m × 380 mm, 2 rolls of 500 sheets

 
140,00 €
   
1 * 2 Roll
 

Multitex® strong, reusable: Particularly tear-resistant viscose fleece wipes, suitable for wet and dry cleaning. The cloths are solvent-resistant and they meet food safety requirements. Multitex® cloths can be washed out after cleaning and reused.

Polytex®, wet-use: Chemical resistant wipes made of 100% polypropylene. Washable and reusable. These tear-proof, fibre-free wipes are a clean and economic solution for many applications.

Multisoft® Cell: White, 4 ply construction, soft and with very high absorptive capacity. For cleaning and polishing applications.

Multiclean® Plus: Highly absorbent wipes with point-to-point imprint. Resistant to solvents, fibre-free and tear-proof. Ideal for use in industrial applications, e.g. for machine maintenance.

White, 4 ply construction, soft and with very high absorptive capacity.

 
  • Packed:
    2 rolls of 500 sheets
  • Colour:
    Blue
  • Description:
    Cleaning cloth Multiclean® plus, blue
  • L×W:
    180 m × 380 mm
  • Seq:
    06