Order Entry
Export
Orders LinkContactUsLinkComponent
PE gloves

PE gloves

For men, L, Transparent, 300 mm, 0,025 mm, 100/bag
Catalog # 112-0061
PE gloves
Catalog # 112-0061
PE gloves
PE gloves

For men, L, Transparent, 300 mm, 0,025 mm, 100/bag

Price
6,00 €
Log in for availability
1 * 100 items
Product Details & Documents

Gloves packed in poly bag. Suitable for use in non hazardous environments.

  • Powder-free
  • Textured for a reliable grip
  • Ambidextrous
  • No toxic admixtures, no heavy metal, free from formaldehyde

Colour: Transparent

Specifications
  • Glove size:
    L
  • Packed:
    100/bag
  • Colour:
    Transparent
  • Length:
    300 mm
  • Thickness:
    0,025 mm
  • Model:
    For men
  • Cat. No.:
    112-0061
  • Disposable / reusable:
    Disposable
  • Powdered / powder-free:
    Powder-free
  • Ambidextrous / hand-specific:
    Ambidextrous