PE gloves
For women, S, Transparent, 280 mm, 0,025 mm, 100/bag
Catalog # 112-0060
Supplier: UNIGLOVES AERTZE UND KLINIKBED
Jump to:
Product Details & Documents
Gloves packed in poly bag. Suitable for use in non hazardous environments.
- Powder-free
- Textured for a reliable grip
- Ambidextrous
- No toxic admixtures, no heavy metal, free from formaldehyde
Colour: Transparent
Specifications
- Glove size:S
- Packed:100/bag
- Colour:Transparent
- Length:280 mm
- Seq:01
- Thickness:0,025 mm
- Model:For women
- Cat. No.:112-0060
- Disposable / reusable:Disposable
- Powdered / powder-free:Powder-free
- Ambidextrous / hand-specific:Ambidextrous



