Orders LinkContactUsLinkComponent
Pierce™ ABC Staining Kits
Pierce™ ABC Staining Kits
Ultra-sensitive ABC peroxidase rabbit IgG staining kit, sufficient for at least 10 ml of ABC staining solution, 1 KIT
Catalog # PIER32054
Pierce™ ABC Staining Kits
Catalog # PIER32054
CAS Number:  
Pierce™ ABC Staining Kits
Pierce™ ABC Staining Kits
Ultra-sensitive ABC peroxidase rabbit IgG staining kit, sufficient for at least 10 ml of ABC staining solution, 1 KIT
CAS Number:  
Price
934,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
ABC Staining Kits include reagents for the avidin-biotin complex (ABC) technique, a highly sensitive method for immunohistochemical detection with biotinylated secondary antibodies.
  • Very low background staining
  • Rapid avidin-biotin interaction
Standard Kits containing only the two essential reagents (avidin and biotinylated HRP) can be used with nearly any species of biotinylated secondary antibody. The Ultra-sensitive ABC Peroxidase Staining Kits enable more dilute primary antibodies to be used while producing equal staining intensity. Ultra-sensitive kits are offered as a standard kit and as complete kits, which contain biotinylated secondary antibodies and appropriate blocking serum for use with mouse or rabbit antibodies.
Documents
Specifications
Specifications
  • Pk:
    1 KIT
  • Description:
    Ultra-sensitive ABC peroxidase rabbit IgG staining kit, sufficient for at least 10 ml of ABC staining solution
Specifications