Pierce™ ABC Staining Kits
Ultra-sensitive ABC peroxidase standard staining kit, sufficient for at least 10 ml of ABC staining solution, 1 KIT
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
ABC Staining Kits include reagents for the avidin-biotin complex (ABC) technique, a highly sensitive method for immunohistochemical detection with biotinylated secondary antibodies.
- Very low background staining
- Rapid avidin-biotin interaction
Standard Kits containing only the two essential reagents (avidin and biotinylated HRP) can be used with nearly any species of biotinylated secondary antibody. The Ultra-sensitive ABC Peroxidase Staining Kits enable more dilute primary antibodies to be used while producing equal staining intensity. Ultra-sensitive kits are offered as a standard kit and as complete kits, which contain biotinylated secondary antibodies and appropriate blocking serum for use with mouse or rabbit antibodies.
Specifications
Specifications
- Pk:1 KIT
- Description:Ultra-sensitive ABC peroxidase standard staining kit, sufficient for at least 10 ml of ABC staining solution